About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:1000 to 1:2000, IF: 1:50 to 1:500
Type:
Antigen: SUZ12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77824-446
- 77824-445
- Antigen Symbol:
- SUZ12
- SUZ12
- Antigen Name:
- Polycomb protein SUZ12 protein (131-305AA)
- Polycomb protein SUZ12 protein (131-305AA)
- Antigen Synonyms:
- ChET 9 protein|CHET9|Chromatin precipitated E2F target 9 protein|JJAZ1|Joined to JAZF1 protein|KIAA0160|Polycomb protein SUZ12|Suppressor of zeste 12 homolog|Suppressor of zeste 12 protein homolog|SUZ12|SUZ12 polycomb repressive complex 2 subunit|SUZ12_HUMAN
- ChET 9 protein|CHET9|Chromatin precipitated E2F target 9 protein|JJAZ1|Joined to JAZF1 protein|KIAA0160|Polycomb protein SUZ12|Suppressor of zeste 12 homolog|Suppressor of zeste 12 protein homolog|SUZ12|SUZ12 polycomb repressive complex 2 subunit|SUZ12_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15022
- Q15022
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice