Anti-HSD17B10 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HSD17B10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-118
- 77835-117
- Antigen Symbol:
- HSD17B10
- HSD17B10
- Antigen Name:
- 3-hydroxyacyl-CoA dehydrogenase type-2 protein (2-261AA)
- 3-hydroxyacyl-CoA dehydrogenase type-2 protein (2-261AA)
- Antigen Synonyms:
- 17 beta hydroxysteroid dehydrogenase 10| 17 beta hydroxysteroid dehydrogenase type 10| 17-beta-HSD 10| 17-beta-hydroxysteroid dehydrogenase 10| 17b HSD10| 3 hydroxy 2 methylbutyryl CoA dehydrogenase| 3 hydroxyacyl CoA dehydrogenase type 2| 3 hydroxyacyl CoA dehydrogenase type II| 3-hydroxy-2-methylbutyryl-CoA dehydrogenase| 3-hydroxyacyl-CoA dehydrogenase type II| 3-hydroxyacyl-CoA dehydrogenase type-2| AB binding alcohol dehydrogenase| ABAD| Ads9| Amyloid beta binding polypeptide| Amyloid beta peptide binding alcohol dehydrogenase| Amyloid beta peptide binding protein| CAMR| DUPXp11.22| Endoplasmic Reticulum Amyloid Binding Protein| Endoplasmic reticulum associated amyloid beta peptide binding protein| Endoplasmic reticulum-associated amyloid beta-peptide-binding protein| ER associated amyloid beta-binding protein| ERAB| HADH 2| HADH2| HCD 2| HCD2| HCD2_HUMAN| Hsd17b10| Hydroxyacyl CoA Dehydrogenase type II| Hydroxyacyl Coenzyme A dehydrogenase type II| Hydroxysteroid (17 beta) dehydrogenase 10| Mental retardation X linked syndromic 11| MHBD| Mitochondrial L3 Hydroxyacyl CoA Dehydrogenase| Mitochondrial ribonuclease P protein 2| Mitochondrial RNase P protein 2| MRPP2| MRX17| SCHAD| SDR5C1| Short chain dehydrogenase/reductase family 5C member 1| Short chain L 3 hydroxyacyl CoA dehydrogenase type 2| Short chain type dehydrogenase/reductase XH98G2| Short-chain type dehydrogenase/reductase XH98G2| Type 10 17b HSD| Type 10 17beta hydroxysteroid dehydrogenase| Type II HADH| XH98G2
- 17 beta hydroxysteroid dehydrogenase 10| 17 beta hydroxysteroid dehydrogenase type 10| 17-beta-HSD 10| 17-beta-hydroxysteroid dehydrogenase 10| 17b HSD10| 3 hydroxy 2 methylbutyryl CoA dehydrogenase| 3 hydroxyacyl CoA dehydrogenase type 2| 3 hydroxyacyl CoA dehydrogenase type II| 3-hydroxy-2-methylbutyryl-CoA dehydrogenase| 3-hydroxyacyl-CoA dehydrogenase type II| 3-hydroxyacyl-CoA dehydrogenase type-2| AB binding alcohol dehydrogenase| ABAD| Ads9| Amyloid beta binding polypeptide| Amyloid beta peptide binding alcohol dehydrogenase| Amyloid beta peptide binding protein| CAMR| DUPXp11.22| Endoplasmic Reticulum Amyloid Binding Protein| Endoplasmic reticulum associated amyloid beta peptide binding protein| Endoplasmic reticulum-associated amyloid beta-peptide-binding protein| ER associated amyloid beta-binding protein| ERAB| HADH 2| HADH2| HCD 2| HCD2| HCD2_HUMAN| Hsd17b10| Hydroxyacyl CoA Dehydrogenase type II| Hydroxyacyl Coenzyme A dehydrogenase type II| Hydroxysteroid (17 beta) dehydrogenase 10| Mental retardation X linked syndromic 11| MHBD| Mitochondrial L3 Hydroxyacyl CoA Dehydrogenase| Mitochondrial ribonuclease P protein 2| Mitochondrial RNase P protein 2| MRPP2| MRX17| SCHAD| SDR5C1| Short chain dehydrogenase/reductase family 5C member 1| Short chain L 3 hydroxyacyl CoA dehydrogenase type 2| Short chain type dehydrogenase/reductase XH98G2| Short-chain type dehydrogenase/reductase XH98G2| Type 10 17b HSD| Type 10 17beta hydroxysteroid dehydrogenase| Type II HADH| XH98G2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99714
- Q99714
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice