Anti-USP46 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:200 to 1:500
Type:
Antigen: USP46
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-149
- 77804-150
- Antigen Symbol:
- USP46
- USP46
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 46 protein (66-166AA)
- Ubiquitin carboxyl-terminal hydrolase 46 protein (66-166AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 46|FLJ11850|FLJ12552|FLJ14283|FLJ39393|Ubiquitin carboxyl terminal hydrolase 46|Ubiquitin carboxyl-terminal hydrolase 46|Ubiquitin specific peptidase 46|Ubiquitin specific processing protease 46|Ubiquitin specific protease 46|Ubiquitin thioesterase 46|Ubiquitin thiolesterase 46|Ubiquitin-specific-processing protease 46|UBP46_HUMAN|USP46
- Deubiquitinating enzyme 46|FLJ11850|FLJ12552|FLJ14283|FLJ39393|Ubiquitin carboxyl terminal hydrolase 46|Ubiquitin carboxyl-terminal hydrolase 46|Ubiquitin specific peptidase 46|Ubiquitin specific processing protease 46|Ubiquitin specific protease 46|Ubiquitin thioesterase 46|Ubiquitin thiolesterase 46|Ubiquitin-specific-processing protease 46|UBP46_HUMAN|USP46
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62068
- P62068
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together