Anti-KLHL12 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: KLHL12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77827-412
- 77827-411
- Antigen Symbol:
- KLHL12
- KLHL12
- Antigen Name:
- Kelch-like protein 12 protein (1-220AA)
- Kelch-like protein 12 protein (1-220AA)
- Antigen Synonyms:
- C3IP1|CUL3 interacting protein 1|CUL3-interacting protein 1|DKIR|DKIR homolog|FLJ27152|hDKIR|Kelch like 12 (Drosophila)|Kelch like 12|Kelch like 12 variant|Kelch like protein 12|Kelch like protein C3IP1|kelch-like family member 12|Kelch-like protein 12|KLH12_HUMAN|KLHL 12|klhl12|OTTHUMP00000038799
- C3IP1|CUL3 interacting protein 1|CUL3-interacting protein 1|DKIR|DKIR homolog|FLJ27152|hDKIR|Kelch like 12 (Drosophila)|Kelch like 12|Kelch like 12 variant|Kelch like protein 12|Kelch like protein C3IP1|kelch-like family member 12|Kelch-like protein 12|KLH12_HUMAN|KLHL 12|klhl12|OTTHUMP00000038799
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53G59
- Q53G59
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice