Anti-PECR Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PECR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-693
- 77793-694
- Antigen Symbol:
- PECR
- PECR
- Antigen Name:
- Peroxisomal trans-2-enoyl-CoA reductase protein (1-303AA)
- Peroxisomal trans-2-enoyl-CoA reductase protein (1-303AA)
- Antigen Synonyms:
- 2 4 dienoyl CoA reductase related protein|2|4-dienoyl-CoA reductase-related protein|DCR RP|DCR-RP|DCRRP|HPDHase|HSA250303|OTTHUMP00000206786|PECR|PECR_HUMAN|Peroxisomal trans 2 enoyl CoA reductase|Peroxisomal trans-2-enoyl-CoA reductase|PRO1004|Putative short chain alcohol dehydrogenase|pVI ARL|pVI-ARL|PVIARL|SDR29C1|Short chain dehydrogenase/reductase family 29C member 1|TERP
- 2 4 dienoyl CoA reductase related protein|2|4-dienoyl-CoA reductase-related protein|DCR RP|DCR-RP|DCRRP|HPDHase|HSA250303|OTTHUMP00000206786|PECR|PECR_HUMAN|Peroxisomal trans 2 enoyl CoA reductase|Peroxisomal trans-2-enoyl-CoA reductase|PRO1004|Putative short chain alcohol dehydrogenase|pVI ARL|pVI-ARL|PVIARL|SDR29C1|Short chain dehydrogenase/reductase family 29C member 1|TERP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BY49
- Q9BY49
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice