Anti-ABHD14A Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ABHD14A
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-134
- 77839-133
- Antigen Symbol:
- ABHD14A
- ABHD14A
- Antigen Name:
- Alpha/beta hydrolase domain-containing protein 14A protein (56-258AA)
- Alpha/beta hydrolase domain-containing protein 14A protein (56-258AA)
- Antigen Synonyms:
- FLJ60317|ABHD14A|ABHEA_HUMAN|Abhydrolase domain containing 14A|Abhydrolase domain containing protein 14A|Abhydrolase domain-containing protein 14A|Alpha/beta hydrolase domain-containing protein 14A|DORZ1|FLJ60467
- FLJ60317|ABHD14A|ABHEA_HUMAN|Abhydrolase domain containing 14A|Abhydrolase domain containing protein 14A|Abhydrolase domain-containing protein 14A|Alpha/beta hydrolase domain-containing protein 14A|DORZ1|FLJ60467
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BUJ0
- Q9BUJ0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice