Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-USP26 Rabbit Recombinant Polyclonal Antibody
Anti-USP26 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-USP26 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77839-207
  • 77839-208
  • Antigen Symbol:
  • USP26
  • USP26
  • Antigen Name:
  • Ubiquitin carboxyl-terminal hydrolase 26 protein (664-913AA)
  • Ubiquitin carboxyl-terminal hydrolase 26 protein (664-913AA)
  • Antigen Synonyms:
  • Deubiquitinating enzyme 26| MGC120066| MGC120067| MGC120068| Ubiquitin carboxyl terminal hydrolase 26| Ubiquitin carboxyl-terminal hydrolase 26| Ubiquitin specific peptidase 26| Ubiquitin specific processing protease 26| Ubiquitin specific protease 26| Ubiquitin thioesterase 26| Ubiquitin thiolesterase 26| Ubiquitin-specific-processing protease 26| UBP26_HUMAN| USP 26| USP26
  • Deubiquitinating enzyme 26| MGC120066| MGC120067| MGC120068| Ubiquitin carboxyl terminal hydrolase 26| Ubiquitin carboxyl-terminal hydrolase 26| Ubiquitin specific peptidase 26| Ubiquitin specific processing protease 26| Ubiquitin specific protease 26| Ubiquitin thioesterase 26| Ubiquitin thiolesterase 26| Ubiquitin-specific-processing protease 26| UBP26_HUMAN| USP 26| USP26
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9BXU7
  • Q9BXU7
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together