Anti-CYP51A1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: CYP51A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77822-767
- 77822-768
- Antigen Symbol:
- CYP51A1
- CYP51A1
- Antigen Name:
- Lanosterol 14-alpha demethylase protein (270-437AA)
- Lanosterol 14-alpha demethylase protein (270-437AA)
- Antigen Synonyms:
- CP51|CP51A_HUMAN|CYP51|CYP51A1|CYPL1|CYPLI|Cytochrome P450 51|Cytochrome P450 51A1|Cytochrome P450 family 51 subfamily A member 1|Cytochrome P450 family 51 subfamily A polypeptide 1|Cytochrome P450|51 (lanosterol 14 alpha demethylase)|Cytochrome P450-14DM|Cytochrome P45014DM|Cytochrome P450LI|Lanosterol 14-alpha demethylase|LDM|P450 14DM|P450L1|Sterol 14-alpha demethylase
- CP51|CP51A_HUMAN|CYP51|CYP51A1|CYPL1|CYPLI|Cytochrome P450 51|Cytochrome P450 51A1|Cytochrome P450 family 51 subfamily A member 1|Cytochrome P450 family 51 subfamily A polypeptide 1|Cytochrome P450|51 (lanosterol 14 alpha demethylase)|Cytochrome P450-14DM|Cytochrome P45014DM|Cytochrome P450LI|Lanosterol 14-alpha demethylase|LDM|P450 14DM|P450L1|Sterol 14-alpha demethylase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16850
- Q16850
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice