Anti-PMP22 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PMP22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-570
- 77794-571
- Antigen Symbol:
- PMP22
- PMP22
- Antigen Name:
- Peripheral myelin protein 22 protein (32-64AA)
- Peripheral myelin protein 22 protein (32-64AA)
- Antigen Synonyms:
- CMT1A|CMT1E|DSS|GAS-3|GAS3|Growth Arrest Specific 3|Growth arrest-specific protein 3|HMSNIA|HNPP|MGC20769|Peripheral myelin protein 22|PMP-22|PMP22|PMP22_HUMAN|Sp110|Trembler
- CMT1A|CMT1E|DSS|GAS-3|GAS3|Growth Arrest Specific 3|Growth arrest-specific protein 3|HMSNIA|HNPP|MGC20769|Peripheral myelin protein 22|PMP-22|PMP22|PMP22_HUMAN|Sp110|Trembler
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q01453
- Q01453
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice