Anti-SIL1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SIL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-369
- 77839-368
- Antigen Symbol:
- SIL1
- SIL1
- Antigen Name:
- Nucleotide exchange factor SIL1 protein (32-174AA)
- Nucleotide exchange factor SIL1 protein (32-174AA)
- Antigen Synonyms:
- BAP|BiP associated protein|BiP-associated protein|Endoplasmic reticulum chaperone SIL 1|Endoplasmic reticulum chaperone SIL1|MSS|Nucleotide exchange factor SIL 1|Nucleotide exchange factor SIL1|SIL 1|sil1|SIL1 homolog|SIL1 homolog endoplasmic reticulum chaperone (S. cerevisiae)|SIL1 homolog endoplasmic reticulum chaperone|SIL1_HUMAN|UGL 5|ULG5
- BAP|BiP associated protein|BiP-associated protein|Endoplasmic reticulum chaperone SIL 1|Endoplasmic reticulum chaperone SIL1|MSS|Nucleotide exchange factor SIL 1|Nucleotide exchange factor SIL1|SIL 1|sil1|SIL1 homolog|SIL1 homolog endoplasmic reticulum chaperone (S. cerevisiae)|SIL1 homolog endoplasmic reticulum chaperone|SIL1_HUMAN|UGL 5|ULG5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H173
- Q9H173
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice