About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: PIK3R6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77828-246
- 77828-247
- Antigen Symbol:
- PIK3R6
- PIK3R6
- Antigen Name:
- Phosphoinositide 3-kinase regulatory subunit 6 protein (566-667AA)
- Phosphoinositide 3-kinase regulatory subunit 6 protein (566-667AA)
- Antigen Synonyms:
- C17orf38|Chromosome 17 open reading frame 38|HsT41028|p84 PI3K adapter protein|p84 PIKAP|p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein|p87 PI3K adapter protein|p87PIKAP|Phosphatidylinositol 3-kinase|regulatory subunit 6|Phosphoinositide 3-kinase gamma adapter protein of 87 kDa|Phosphoinositide 3-kinase regulatory subunit 6|PI3Kgamma adapter protein of 87 kDa|PI3R6_HUMAN|PIK3R6
- C17orf38|Chromosome 17 open reading frame 38|HsT41028|p84 PI3K adapter protein|p84 PIKAP|p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein|p87 PI3K adapter protein|p87PIKAP|Phosphatidylinositol 3-kinase|regulatory subunit 6|Phosphoinositide 3-kinase gamma adapter protein of 87 kDa|Phosphoinositide 3-kinase regulatory subunit 6|PI3Kgamma adapter protein of 87 kDa|PI3R6_HUMAN|PIK3R6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5UE93
- Q5UE93
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice