Anti-USP17L5 Rabbit Recombinant Polyclonal Antibody
Catalog # 77818-247
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 17-like protein 5 protein (19-235AA)
- Antigen Symbol:USP17L5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:A8MUK1
- Antigen Synonyms:USP17L5|Ubiquitin carboxyl-terminal hydrolase 17-like protein 5|EC 3.4.19.12|Deubiquitinating enzyme 17-like protein 5|Ubiquitin thioesterase 17-like protein 5|Ubiquitin-specific-processing protease 17-like protein 5
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:200 to 1:500
Type:
Antigen: USP17L5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human