Anti-HSD17B10 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HSD17B10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-115
- 77835-116
- Antigen Symbol:
- 17 beta hydroxysteroid dehydrogenase 10;17 beta hydroxysteroid dehydrogenase type 10;17-beta-HSD 10;17-beta-hydroxysteroid dehydrogenase 10;17b HSD10;3 hydroxy 2 methylbutyryl CoA dehydrogenase;3 hydroxyacyl CoA dehydrogenase type 2;3 hydroxyacyl CoA dehydrogenase type II;3-hydroxy-2-methylbutyryl-CoA dehydrogenase;3-hydroxyacyl-CoA dehydrogenase type II;3-hydroxyacyl-CoA dehydrogenase type-2;AB binding alcohol dehydrogenase;ABAD;Ads9;Amyloid beta binding polypeptide;Amyloid beta peptide binding alcohol dehydrogenase;Amyloid beta peptide binding protein;CAMR;DUPXp11.22;Endoplasmic Reticulum Amyloid Binding Protein;Endoplasmic reticulum associated amyloid beta peptide binding protein;Endoplasmic reticulum-associated amyloid beta-peptide-binding protein;ER associated amyloid beta-binding protein;ERAB;HADH 2;HADH2;HCD 2;HCD2;HCD2_HUMAN;Hsd17b10;Hydroxyacyl CoA Dehydrogenase type II;Hydroxyacyl Coenzyme A dehydrogenase type II;Hydroxysteroid (17 beta) dehydrogenase 10;Mental retardation X linked syndromic 11;MHBD;Mitochondrial L3 Hydroxyacyl CoA Dehydrogenase;Mitochondrial ribonuclease P protein 2;Mitochondrial RNase P protein 2;MRPP2;MRX17;SCHAD;SDR5C1;Short chain dehydrogenase/reductase family 5C member 1;Short chain L 3 hydroxyacyl CoA dehydrogenase type 2;Short chain type dehydrogenase/reductase XH98G2;Short-chain type dehydrogenase/reductase XH98G2;Type 10 17b HSD;Type 10 17beta hydroxysteroid dehydrogenase;Type II HADH;XH98G2
- 17 beta hydroxysteroid dehydrogenase 10;17 beta hydroxysteroid dehydrogenase type 10;17-beta-HSD 10;17-beta-hydroxysteroid dehydrogenase 10;17b HSD10;3 hydroxy 2 methylbutyryl CoA dehydrogenase;3 hydroxyacyl CoA dehydrogenase type 2;3 hydroxyacyl CoA dehydrogenase type II;3-hydroxy-2-methylbutyryl-CoA dehydrogenase;3-hydroxyacyl-CoA dehydrogenase type II;3-hydroxyacyl-CoA dehydrogenase type-2;AB binding alcohol dehydrogenase;ABAD;Ads9;Amyloid beta binding polypeptide;Amyloid beta peptide binding alcohol dehydrogenase;Amyloid beta peptide binding protein;CAMR;DUPXp11.22;Endoplasmic Reticulum Amyloid Binding Protein;Endoplasmic reticulum associated amyloid beta peptide binding protein;Endoplasmic reticulum-associated amyloid beta-peptide-binding protein;ER associated amyloid beta-binding protein;ERAB;HADH 2;HADH2;HCD 2;HCD2;HCD2_HUMAN;Hsd17b10;Hydroxyacyl CoA Dehydrogenase type II;Hydroxyacyl Coenzyme A dehydrogenase type II;Hydroxysteroid (17 beta) dehydrogenase 10;Mental retardation X linked syndromic 11;MHBD;Mitochondrial L3 Hydroxyacyl CoA Dehydrogenase;Mitochondrial ribonuclease P protein 2;Mitochondrial RNase P protein 2;MRPP2;MRX17;SCHAD;SDR5C1;Short chain dehydrogenase/reductase family 5C member 1;Short chain L 3 hydroxyacyl CoA dehydrogenase type 2;Short chain type dehydrogenase/reductase XH98G2;Short-chain type dehydrogenase/reductase XH98G2;Type 10 17b HSD;Type 10 17beta hydroxysteroid dehydrogenase;Type II HADH;XH98G2
- Antigen Name:
- 3-hydroxyacyl-CoA dehydrogenase type-2 protein (2-261AA)
- 3-hydroxyacyl-CoA dehydrogenase type-2 protein (2-261AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99714
- Q99714
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice