Anti-ARID3A Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ARID3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77834-610
- 77834-609
- Antigen Symbol:
- ARID3A
- ARID3A
- Antigen Name:
- AT-rich interactive domain-containing protein 3A protein (26-137AA)
- AT-rich interactive domain-containing protein 3A protein (26-137AA)
- Antigen Synonyms:
- ARI3A_HUMAN|ARID domain containing 3A|ARID domain-containing protein 3A|ARID3A|AT rich interactive domain 3A (BRIGHT- like) protein|AT rich interactive domain 3A (BRIGHT-like)|AT rich interactive domain-containing protein 3A|AT-rich interactive domain-containing protein 3A|B cell regulator of IgH transcription|B-cell regulator of IgH transcription|Bright|dead ringer like 1|dead ringer|Drosophila|homolog-like 1|Dead ringer-like protein 1|DRIL1|DRIL3|DRX|E2F binding protein 1|E2F-binding protein 1|E2FBP1|Homo sapiens AT rich interactive domain 3A (BRIGHT-like)
- ARI3A_HUMAN|ARID domain containing 3A|ARID domain-containing protein 3A|ARID3A|AT rich interactive domain 3A (BRIGHT- like) protein|AT rich interactive domain 3A (BRIGHT-like)|AT rich interactive domain-containing protein 3A|AT-rich interactive domain-containing protein 3A|B cell regulator of IgH transcription|B-cell regulator of IgH transcription|Bright|dead ringer like 1|dead ringer|Drosophila|homolog-like 1|Dead ringer-like protein 1|DRIL1|DRIL3|DRX|E2F binding protein 1|E2F-binding protein 1|E2FBP1|Homo sapiens AT rich interactive domain 3A (BRIGHT-like)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99856
- Q99856
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice