Anti-STOM Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: STOM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77801-393
- 77801-394
- Antigen Symbol:
- STOM
- STOM
- Antigen Name:
- Erythrocyte band 7 integral membrane protein (55-288AA)
- Erythrocyte band 7 integral membrane protein (55-288AA)
- Antigen Synonyms:
- BND7|EPB7|EPB72|Erythrocyte band 7 integral membrane protein|Erythrocyte membrane protein band 7.2 (stomatin)|Erythrocyte surface protein band 7.2|Protein 7.2b|STOM|STOM_HUMAN|Stomatin
- BND7|EPB7|EPB72|Erythrocyte band 7 integral membrane protein|Erythrocyte membrane protein band 7.2 (stomatin)|Erythrocyte surface protein band 7.2|Protein 7.2b|STOM|STOM_HUMAN|Stomatin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P27105
- P27105
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice