Anti-MTR Rabbit Recombinant Polyclonal Antibody
Catalog # 77834-944
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Methionine synthase protein (923-1265AA)
- Antigen Symbol:MTR
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q99707
- Antigen Synonyms:MTRMethionine synthase|EC 2.1.1.13|5-methyltetrahydrofolate--homocysteine methyltransferase|Vitamin-B12 dependent methionine synthase|MS
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200, IP:1:200 to 1:2000
Type:
Antigen: MTR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human