Anti-C1QTNF9B Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: C1QTNF9B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77818-768
- 77818-769
- Antigen Symbol:
- C1QTNF9B
- C1QTNF9B
- Antigen Name:
- Complement C1q and tumor necrosis factor-related protein 9B protein (2-192AA)
- Complement C1q and tumor necrosis factor-related protein 9B protein (2-192AA)
- Antigen Synonyms:
- C1q/TNF-related protein 9B|C1QTNF9B|C1T9B_HUMAN|Complement C1q and tumor necrosis factor-related protein 9-like|Complement C1q and tumor necrosis factor-related protein 9B|CTRP9B
- C1q/TNF-related protein 9B|C1QTNF9B|C1T9B_HUMAN|Complement C1q and tumor necrosis factor-related protein 9-like|Complement C1q and tumor necrosis factor-related protein 9B|CTRP9B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- B2RNN3
- B2RNN3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice