Anti-MRPL36 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRPL36
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-556
- 77842-557
- Antigen Symbol:
- MRPL36
- MRPL36
- Antigen Name:
- 39S ribosomal protein L36, mitochondrial protein (2-103AA)
- 39S ribosomal protein L36, mitochondrial protein (2-103AA)
- Antigen Synonyms:
- 39S ribosomal protein L36|39S ribosomal protein L36 mitochondrial|BRCA1-interacting protein 1|BRIP1|L36mt|mitochondrial|Mitochondrial ribosomal protein L36|MRP L36|MRP-L36|Mrpl36|PRPL36|Putative BRCA1 interacting protein|RM36_HUMAN|RPMJ
- 39S ribosomal protein L36|39S ribosomal protein L36 mitochondrial|BRCA1-interacting protein 1|BRIP1|L36mt|mitochondrial|Mitochondrial ribosomal protein L36|MRP L36|MRP-L36|Mrpl36|PRPL36|Putative BRCA1 interacting protein|RM36_HUMAN|RPMJ
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P0J6
- Q9P0J6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice