Anti-SLC34A3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC34A3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77833-282
- 77833-281
- Antigen Symbol:
- SLC34A3
- SLC34A3
- Antigen Name:
- Sodium-dependent phosphate transport protein 2C protein (1-76AA)
- Sodium-dependent phosphate transport protein 2C protein (1-76AA)
- Antigen Synonyms:
- HHRH|Na(+) dependent phosphate cotransporter 2C|Na(+) Pi cotransporter 2C|Na(+)-dependent phosphate cotransporter 2C|Na(+)/Pi cotransporter 2C|NaPi 2c|NaPi-2c|NPT2C|NPT2C_HUMAN|NPTIIC|SLC34A3|Sodium dependent phosphate transport protein 2C|Sodium inorganic phosphate cotransporter IIC|Sodium phosphate cotransporter 2C|Sodium phosphate transport protein 2C|Sodium-dependent phosphate transport protein 2C|Sodium-phosphate transport protein 2C|Sodium/inorganic phosphate cotransporter IIC|Sodium/inorganic phosphate cotransporter|type IIC|Sodium/phosphate cotransporter 2C|solute carrier family 34 (sodium phosphate) member 3|solute carrier family 34 (sodium/phosphate contransporter)|member 3|solute carrier family 34 (type II sodium/phosphate contransporter)|Solute carrier family 34 member 3|type IIc Na+ Pi cotransporter|Type IIc Na+/Pi cotransporter
- HHRH|Na(+) dependent phosphate cotransporter 2C|Na(+) Pi cotransporter 2C|Na(+)-dependent phosphate cotransporter 2C|Na(+)/Pi cotransporter 2C|NaPi 2c|NaPi-2c|NPT2C|NPT2C_HUMAN|NPTIIC|SLC34A3|Sodium dependent phosphate transport protein 2C|Sodium inorganic phosphate cotransporter IIC|Sodium phosphate cotransporter 2C|Sodium phosphate transport protein 2C|Sodium-dependent phosphate transport protein 2C|Sodium-phosphate transport protein 2C|Sodium/inorganic phosphate cotransporter IIC|Sodium/inorganic phosphate cotransporter|type IIC|Sodium/phosphate cotransporter 2C|solute carrier family 34 (sodium phosphate) member 3|solute carrier family 34 (sodium/phosphate contransporter)|member 3|solute carrier family 34 (type II sodium/phosphate contransporter)|Solute carrier family 34 member 3|type IIc Na+ Pi cotransporter|Type IIc Na+/Pi cotransporter
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N130
- Q8N130
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice