Anti-HLA-F Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HLA-F
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77816-100
- 77816-101
- Antigen Symbol:
- HLA-F
- HLA-F
- Antigen Name:
- HLA class I histocompatibility antigen, alpha chain F protein (194-303AA)
- HLA class I histocompatibility antigen, alpha chain F protein (194-303AA)
- Antigen Synonyms:
- CDA12|HLA 5.4|HLA class I histocompatibility antigen|alpha chain F|HLA class I molecule|HLA F antigen|HLA-CDA12|HLA-F|HLAF|HLAF_HUMAN|Leukocyte antigen F|MHC class I antigen F
- CDA12|HLA 5.4|HLA class I histocompatibility antigen|alpha chain F|HLA class I molecule|HLA F antigen|HLA-CDA12|HLA-F|HLAF|HLAF_HUMAN|Leukocyte antigen F|MHC class I antigen F
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P30511
- P30511
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice