Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-MTR Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
 
undefined
Anti-MTR Rabbit Recombinant Polyclonal Antibody (FITC)
New Product

 

  • Catalog No:
  • 77834-947
  • 77834-948
  • Antigen Symbol:
  • MTR
  • MTR
  • Antigen Name:
  • Methionine synthase protein (923-1265AA)
  • Methionine synthase protein (923-1265AA)
  • Antigen Synonyms:
  • MTRMethionine synthase|EC 2.1.1.13|5-methyltetrahydrofolate--homocysteine methyltransferase|Vitamin-B12 dependent methionine synthase|MS
  • MTRMethionine synthase|EC 2.1.1.13|5-methyltetrahydrofolate--homocysteine methyltransferase|Vitamin-B12 dependent methionine synthase|MS
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q99707
  • Q99707
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, protein G purified
  • >95%, protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice