Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-USP8 Rabbit Recombinant Polyclonal Antibody
Anti-USP8 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-USP8 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77804-176
  • 77804-177
  • Antigen Symbol:
  • USP8
  • USP8
  • Antigen Name:
  • Ubiquitin carboxyl-terminal hydrolase 8 protein (1-270AA)
  • Ubiquitin carboxyl-terminal hydrolase 8 protein (1-270AA)
  • Antigen Synonyms:
  • Deubiquitinating enzyme 8|FLJ34456|hUBPy|HumORF8|KIAA0055|MGC129718|SPG59|Ubiquitin carboxyl-terminal hydrolase 8|Ubiquitin isopeptidase Y|ubiquitin specific peptidase 8|Ubiquitin thioesterase 8|Ubiquitin-specific-processing protease 8|UBP8_HUMAN|UBPY|Usp8
  • Deubiquitinating enzyme 8|FLJ34456|hUBPy|HumORF8|KIAA0055|MGC129718|SPG59|Ubiquitin carboxyl-terminal hydrolase 8|Ubiquitin isopeptidase Y|ubiquitin specific peptidase 8|Ubiquitin thioesterase 8|Ubiquitin-specific-processing protease 8|UBP8_HUMAN|UBPY|Usp8
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P40818
  • P40818
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together