Anti-SMAD7 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SMAD7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-271
- 77800-270
- Antigen Symbol:
- SMAD7
- SMAD7
- Antigen Name:
- Mothers against decapentaplegic homolog 7 protein (36-247AA)
- Mothers against decapentaplegic homolog 7 protein (36-247AA)
- Antigen Synonyms:
- CRCS3| FLJ16482| hSMAD 7| hSMAD7| MAD (mothers against decapentaplegic Drosophila) homolog 7| MAD| Mad homolog 7| MAD homolog 8| MAD mothers against decapentaplegic homolog 7| MADH 7| MADH 8| MADH6| MADH7| MADH8| Mothers Against Decapentaplegic Drosophila Homolog of 6| Mothers Against Decapentaplegic Drosophila Homolog of 7| Mothers against decapentaplegic homolog 7| Mothers against decapentaplegic homolog 8| Mothers against DPP homolog 7| Mothers against DPP homolog 8| SMA- AND MAD-RELATED PROTEIN 7| SMAD 7| SMAD| SMAD family member 7| SMAD, mothers against DPP homolog 7 (Drosophila)| SMAD, mothers against DPP homolog 7| SMAD6| Smad7| SMAD7_HUMAN
- CRCS3| FLJ16482| hSMAD 7| hSMAD7| MAD (mothers against decapentaplegic Drosophila) homolog 7| MAD| Mad homolog 7| MAD homolog 8| MAD mothers against decapentaplegic homolog 7| MADH 7| MADH 8| MADH6| MADH7| MADH8| Mothers Against Decapentaplegic Drosophila Homolog of 6| Mothers Against Decapentaplegic Drosophila Homolog of 7| Mothers against decapentaplegic homolog 7| Mothers against decapentaplegic homolog 8| Mothers against DPP homolog 7| Mothers against DPP homolog 8| SMA- AND MAD-RELATED PROTEIN 7| SMAD 7| SMAD| SMAD family member 7| SMAD, mothers against DPP homolog 7 (Drosophila)| SMAD, mothers against DPP homolog 7| SMAD6| Smad7| SMAD7_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15105
- O15105
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice