Anti-MBD1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: MBD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-591
- 77841-592
- Antigen Symbol:
- MBD1
- MBD1
- Antigen Name:
- Methyl-CpG-binding domain protein 1 protein (106-355AA)
- Methyl-CpG-binding domain protein 1 protein (106-355AA)
- Antigen Synonyms:
- CXXC 3| CXXC-type zinc finger protein 3| CXXC3| MBD 1| Mbd1| _HUMAN| MECP1 COMPLEX| Methyl CpG binding domain protein 1| Methyl CpG binding domain protein 1 isoform PCM1| Methyl CpG binding protein | Methyl CpG binding protein splice variant 1| Methyl CpG binding protein splice variant 2| Methyl CpG binding protein splice variant 3| Methyl CpG binding protein splice variant 4| Methyl-CpG-binding domain protein 1| Methyl-CpG-binding protein | PCM 1| PCM1| Protein containing methyl-CpG-binding domain 1| Regulator of fibroblast growth factor 2 (FGF 2) transcription| RFT| The regulator of fibroblast growth factor 2(FGF 2) transcription
- CXXC 3| CXXC-type zinc finger protein 3| CXXC3| MBD 1| Mbd1| _HUMAN| MECP1 COMPLEX| Methyl CpG binding domain protein 1| Methyl CpG binding domain protein 1 isoform PCM1| Methyl CpG binding protein | Methyl CpG binding protein splice variant 1| Methyl CpG binding protein splice variant 2| Methyl CpG binding protein splice variant 3| Methyl CpG binding protein splice variant 4| Methyl-CpG-binding domain protein 1| Methyl-CpG-binding protein | PCM 1| PCM1| Protein containing methyl-CpG-binding domain 1| Regulator of fibroblast growth factor 2 (FGF 2) transcription| RFT| The regulator of fibroblast growth factor 2(FGF 2) transcription
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UIS9
- Q9UIS9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice