Anti-GAS8 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: GAS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77821-284
- 77821-283
- Antigen Symbol:
- GAS8
- GAS8
- Antigen Name:
- Synthetic peptide of GAS8
- Synthetic peptide of GAS8
- Antigen Synonyms:
- Growth arrest specific protein 8| MGC138326|GAS 11|GAS 8|GAS-11|GAS-8|GAS11|Gas8|GAS8_HUMAN|Growth arrest specific 11|Growth arrest specific 8|Growth arrest specific protein 11|Growth arrest-specific protein 11|Growth arrest-specific protein 8
- Growth arrest specific protein 8| MGC138326|GAS 11|GAS 8|GAS-11|GAS-8|GAS11|Gas8|GAS8_HUMAN|Growth arrest specific 11|Growth arrest specific 8|Growth arrest specific protein 11|Growth arrest-specific protein 11|Growth arrest-specific protein 8
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95995
- O95995
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice