Anti-CTNNBIP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CTNNBIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-759
- 77838-760
- Antigen Symbol:
- CTNNBIP1
- CTNNBIP1
- Antigen Name:
- Beta-catenin-interacting protein 1 protein (1-81AA)
- Beta-catenin-interacting protein 1 protein (1-81AA)
- Antigen Synonyms:
- Beta catenin interacting protein 1|Beta catenin interacting protein ICAT|Beta-catenin-interacting protein 1|Catenin beta interacting protein 1|CNBP1_HUMAN|CTNNBIP 1|CTNNBIP1|ICAT|Inhibitor of beta catenin and Tcf 4|Inhibitor of beta-catenin and Tcf-4|MGC15093
- Beta catenin interacting protein 1|Beta catenin interacting protein ICAT|Beta-catenin-interacting protein 1|Catenin beta interacting protein 1|CNBP1_HUMAN|CTNNBIP 1|CTNNBIP1|ICAT|Inhibitor of beta catenin and Tcf 4|Inhibitor of beta-catenin and Tcf-4|MGC15093
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NSA3
- Q9NSA3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice