Anti-AK3 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AK3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-563
- 77841-564
- Antigen Symbol:
- AK3
- AK3
- Antigen Name:
- GTP:AMP phosphotransferase AK3, mitochondrial protein (142-227AA)
- GTP:AMP phosphotransferase AK3, mitochondrial protein (142-227AA)
- Antigen Synonyms:
- Adenylate kinase 3 alpha-like 1|Adenylate kinase 3|Adenylate kinase 3, formerly|adenylate kinase 6, adenylate kinase 3 like 1|AK 3|AK3|AK3, formerly|AK3L1|AK4|AK4, mouse, homolog of|AK6|AKL3L|AKL3L1|FIX|GTP:AMP phosphotransferase|GTP:AMP phosphotransferase, mitochondrial|KAD3_HUMAN|mitochondrial|OTTHUMP00000021015|OTTHUMP00000021016|RP11 6J24.4
- Adenylate kinase 3 alpha-like 1|Adenylate kinase 3|Adenylate kinase 3, formerly|adenylate kinase 6, adenylate kinase 3 like 1|AK 3|AK3|AK3, formerly|AK3L1|AK4|AK4, mouse, homolog of|AK6|AKL3L|AKL3L1|FIX|GTP:AMP phosphotransferase|GTP:AMP phosphotransferase, mitochondrial|KAD3_HUMAN|mitochondrial|OTTHUMP00000021015|OTTHUMP00000021016|RP11 6J24.4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UIJ7
- Q9UIJ7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together