Anti-DUSP15 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DUSP15
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-478
- 77835-477
- Antigen Symbol:
- DUSP15
- DUSP15
- Antigen Name:
- Dual specificity protein phosphatase 15 protein (1-295AA)
- Dual specificity protein phosphatase 15 protein (1-295AA)
- Antigen Synonyms:
- bA243J16.5|Chromosome 20 open reading frame 57|Dual specificity phosphatase 15|Dual specificity phosphatase like 15|Dual specificity protein phosphatase 15|DUS15_HUMAN|DUSP15|EC 3.1.3.16|EC 3.1.3.48|FLJ20645|Vaccinia virus VH1 related dual specific protein phosphatase Y|Vaccinia virus VH1-related dual-specific protein phosphatase Y|VH1 related member Y|VH1-related member Y|VHY
- bA243J16.5|Chromosome 20 open reading frame 57|Dual specificity phosphatase 15|Dual specificity phosphatase like 15|Dual specificity protein phosphatase 15|DUS15_HUMAN|DUSP15|EC 3.1.3.16|EC 3.1.3.48|FLJ20645|Vaccinia virus VH1 related dual specific protein phosphatase Y|Vaccinia virus VH1-related dual-specific protein phosphatase Y|VH1 related member Y|VH1-related member Y|VHY
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H1R2
- Q9H1R2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice