About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: DPYSL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77807-729
- 77807-730
- Antigen Symbol:
- DPYSL3
- DPYSL3
- Antigen Name:
- Fusion protein of DPYSL3
- Fusion protein of DPYSL3
- Antigen Synonyms:
- Collapsin response mediator protein 4|Collapsin response mediator protein 4 long|CRMP 4|CRMP-4|CRMP4|Dihydropyrimidinase like 3|Dihydropyrimidinase related protein 3|Dihydropyrimidinase-related protein 3|DPYL3_HUMAN|DRP 3|DRP-3|DRP3|LCRMP|TUC4|ULIP 1|ULIP|ULIP-1|ULIP1|UNc 33 like phosphoprotein 1|Unc 33 like phosphoprotein|Unc-33-like phosphoprotein 1
- Collapsin response mediator protein 4|Collapsin response mediator protein 4 long|CRMP 4|CRMP-4|CRMP4|Dihydropyrimidinase like 3|Dihydropyrimidinase related protein 3|Dihydropyrimidinase-related protein 3|DPYL3_HUMAN|DRP 3|DRP-3|DRP3|LCRMP|TUC4|ULIP 1|ULIP|ULIP-1|ULIP1|UNc 33 like phosphoprotein 1|Unc 33 like phosphoprotein|Unc-33-like phosphoprotein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14195
- Q14195
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice