Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SSNA1 Rabbit Recombinant Polyclonal Antibody
Anti-SSNA1 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-SSNA1 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77801-107
  • 77801-106
  • Antigen Symbol:
  • SSNA1
  • SSNA1
  • Antigen Name:
  • Sjoegren syndrome nuclear autoantigen 1 protein (1-119AA)
  • Sjoegren syndrome nuclear autoantigen 1 protein (1-119AA)
  • Antigen Synonyms:
  • SSNA1|NA14Sjoegren syndrome nuclear autoantigen 1|Nuclear autoantigen of 14 kDa
  • SSNA1|NA14Sjoegren syndrome nuclear autoantigen 1|Nuclear autoantigen of 14 kDa
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O43805
  • O43805
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • Storage Temperature:
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications