Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MBD1 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Anti-MBD1 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MBD1 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77841-595
  • 77841-596
  • Antigen Symbol:
  • MBD1
  • MBD1
  • Antigen Name:
  • Methyl-CpG-binding domain protein 1 protein (106-355AA)
  • Methyl-CpG-binding domain protein 1 protein (106-355AA)
  • Antigen Synonyms:
  • CXXC 3|CXXC-type zinc finger protein 3|CXXC3|MBD 1|Mbd1|MBD1_HUMAN|MECP1 COMPLEX|Methyl CpG binding domain protein 1|Methyl CpG binding domain protein 1 isoform PCM1|Methyl CpG binding protein MBD1|Methyl CpG binding protein splice variant 1|Methyl CpG binding protein splice variant 2|Methyl CpG binding protein splice variant 3|Methyl CpG binding protein splice variant 4|Methyl-CpG-binding domain protein 1|Methyl-CpG-binding protein MBD1|PCM 1|PCM1|Protein containing methyl-CpG-binding domain 1|Regulator of fibroblast growth factor 2 (FGF 2) transcription|RFT|The regulator of fibroblast growth factor 2(FGF 2) transcription
  • CXXC 3|CXXC-type zinc finger protein 3|CXXC3|MBD 1|Mbd1|MBD1_HUMAN|MECP1 COMPLEX|Methyl CpG binding domain protein 1|Methyl CpG binding domain protein 1 isoform PCM1|Methyl CpG binding protein MBD1|Methyl CpG binding protein splice variant 1|Methyl CpG binding protein splice variant 2|Methyl CpG binding protein splice variant 3|Methyl CpG binding protein splice variant 4|Methyl-CpG-binding domain protein 1|Methyl-CpG-binding protein MBD1|PCM 1|PCM1|Protein containing methyl-CpG-binding domain 1|Regulator of fibroblast growth factor 2 (FGF 2) transcription|RFT|The regulator of fibroblast growth factor 2(FGF 2) transcription
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UIS9
  • Q9UIS9
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications