Anti-TERF2IP Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TERF2IP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77839-985
- 77839-984
- Antigen Symbol:
- TERF2IP
- TERF2IP
- Antigen Name:
- Telomeric repeat-binding factor 2-interacting protein 1 protein (1-280AA)
- Telomeric repeat-binding factor 2-interacting protein 1 protein (1-280AA)
- Antigen Synonyms:
- Dopamine receptor interacting protein 5|Dopamine receptor-interacting protein 5|DRIP 5|DRIP5|hRap1|MGC105533|RAP 1|RAP1 homolog|RAP1, yeast, homolog if|RAP1, yeast, homolog of|Repressor/activator protein 1 homolog|TE2IP_HUMAN|Telomeric Repeat Binding Factor 2 Interacting Protein|Telomeric repeat-binding factor 2-interacting protein 1|TERF2-interacting protein|TERF2-interacting telomeric protein 1|TERF2IP|TRF2 Interacting Telomeric Protein RAP1|TRF2 interacting telomeric RAP1 protein|TRF2-interacting telomeric protein 1|TRF2-interacting telomeric protein
- Dopamine receptor interacting protein 5|Dopamine receptor-interacting protein 5|DRIP 5|DRIP5|hRap1|MGC105533|RAP 1|RAP1 homolog|RAP1, yeast, homolog if|RAP1, yeast, homolog of|Repressor/activator protein 1 homolog|TE2IP_HUMAN|Telomeric Repeat Binding Factor 2 Interacting Protein|Telomeric repeat-binding factor 2-interacting protein 1|TERF2-interacting protein|TERF2-interacting telomeric protein 1|TERF2IP|TRF2 Interacting Telomeric Protein RAP1|TRF2 interacting telomeric RAP1 protein|TRF2-interacting telomeric protein 1|TRF2-interacting telomeric protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NYB0
- Q9NYB0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice