Anti-CREB3L1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77833-945
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Cyclic AMP-responsive element-binding protein 3-like protein 1 protein (396-500AA)
- Antigen Symbol:CREB3L1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96BA8
- Antigen Synonyms:BBF-2 homolog|CAMP responsive element binding protein 3 like 1|cAMP-responsive element-binding protein 3-like protein 1|CR3L1_HUMAN|Creb3l1|Cyclic AMP responsive element binding protein 3 like protein 1|OASIS|Old astrocyte specifically-induced substance|Processed cyclic AMP-responsive element-binding protein 3-like protein 1
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:200 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CREB3L1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat