Anti-PRKAA1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77823-386
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:5\\\'-AMP-activated protein kinase catalytic subunit alpha-1 protein (325-543AA)
- Antigen Symbol:PRKAA1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 μg
- Form:Liquid
- Gene ID:Q13131
- Antigen Synonyms:5 AMP activated protein kinase alpha 1catalytic subunit|5 AMP activated protein kinase catalytic alpha 1 chain|5' AMP activated protein kinase catalytic subunit alpha 1|5'-AMP-activated protein kinase catalytic subunit alpha-1|AAPK1|AAPK1_HUMAN|ACACA kinase|acetyl CoA carboxylase kinase|AI194361|AI450832|AL024255|AMP -activate kinase alpha 1 subunit|AMP-activate kinase alpha 1 subunit |AMP-activated protein kinase, catalytic, alpha -1|AMPK 1|AMPK alpha 1|AMPK alpha 1 chain|AMPK|AMPK subunit alpha 1|AMPK subunit alpha-1|AMPK1|AMPKa1|AMPKalpha1|C130083N04Rik|cb116|EC 2.7.11.1|HMG CoA reductase kinase|HMGCR kinase|hormone sensitive lipase kinase|Hydroxymethylglutaryl CoA reductase kinase|im:7154392|kinase AMPK alpha1|MGC33776|MGC57364|OTTHUMP00000161795|OTTHUMP00000161796|PRKAA 1||Protein kinase AMP activated alpha 1 catalytic subunit|SNF1-like protein AMPK|SNF1A|Tau protein kinase |wu:fa94c10
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: PRKAA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse