Anti-tar Rabbit Recombinant Polyclonal Antibody
77817-046
New Product
- Antigen Name:Recombinant Escherichia coli Methyl-accepting chemotaxis protein II protein (212-553AA)
- Antigen Symbol:tar
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:E. coli
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P07017
- Antigen Synonyms:tar| cheM| b1886| JW1875Methyl-accepting chemotaxis protein II| MCP-II| Aspartate chemoreceptor protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: tar
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli