Anti-ACS8 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: ACS8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
- Catalog No:
- 77837-840
- 77837-839
- Antigen Symbol:
- ACS8
- ACS8
- Antigen Name:
- Recombinant Arabidopsis thaliana 1-aminocyclopropane-1-carboxylate synthase 8 protein (1-469AA)
- Recombinant Arabidopsis thaliana 1-aminocyclopropane-1-carboxylate synthase 8 protein (1-469AA)
- Antigen Synonyms:
- ACS8|At4g37770|T28I19.501-aminocyclopropane-1-carboxylate synthase 8|ACC synthase 8|EC 4.4.1.14|S-adenosyl-L-methionine methylthioadenosine-lyase 8
- ACS8|At4g37770|T28I19.501-aminocyclopropane-1-carboxylate synthase 8|ACC synthase 8|EC 4.4.1.14|S-adenosyl-L-methionine methylthioadenosine-lyase 8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- A. thaliana
- A. thaliana
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9T065
- Q9T065
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice