Anti-QDPR Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: QDPR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-282
- 77796-281
- Antigen Symbol:
- QDPR
- QDPR
- Antigen Name:
- Dihydropteridine reductase protein (2-244AA)
- Dihydropteridine reductase protein (2-244AA)
- Antigen Synonyms:
- 6,7 dihydropteridine reductase|DHPR|DHPR_HUMAN|Dihydropteridine reductase|HDHPR|HPR|PKU2|Qdpr|Quinoid dihydropteridine reductase|SDR33C1|Short chain dehydrogenase/reductase family 33C, member 1
- 6,7 dihydropteridine reductase|DHPR|DHPR_HUMAN|Dihydropteridine reductase|HDHPR|HPR|PKU2|Qdpr|Quinoid dihydropteridine reductase|SDR33C1|Short chain dehydrogenase/reductase family 33C, member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P09417
- P09417
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice