Anti-USP36 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP36
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-097
- 77840-098
- Antigen Symbol:
- USP36
- USP36
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 36 protein (670-881AA)
- Ubiquitin carboxyl-terminal hydrolase 36 protein (670-881AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 1|Deubiquitinating enzyme 36|DUB1|FLJ12851|KIAA1453|Ubiquitin carboxyl-terminal hydrolase 36|Ubiquitin specific peptidase 36|Ubiquitin specific processing protease 36|Ubiquitin thioesterase 36|Ubiquitin thiolesterase 36|Ubiquitin-specific-processing protease 36|UBP36_HUMAN|Usp36
- Deubiquitinating enzyme 1|Deubiquitinating enzyme 36|DUB1|FLJ12851|KIAA1453|Ubiquitin carboxyl-terminal hydrolase 36|Ubiquitin specific peptidase 36|Ubiquitin specific processing protease 36|Ubiquitin thioesterase 36|Ubiquitin thiolesterase 36|Ubiquitin-specific-processing protease 36|UBP36_HUMAN|Usp36
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P275
- Q9P275
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together