Anti-REST Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: REST
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-058
- 77797-057
- Antigen Symbol:
- REST
- REST
- Antigen Name:
- RE1-silencing transcription factor protein (250-350AA)
- RE1-silencing transcription factor protein (250-350AA)
- Antigen Synonyms:
- Neural Restrictive Silencer Factor|Neural-restrictive silencer factor|Neuron Restrictive Silencer Factor|NRSF|RE1 silencing transcription factor|RE1-silencing transcription factor|Repressor binding to the X2 box|rest|REST protein|REST_HUMAN|X2 box repressor|XBR
- Neural Restrictive Silencer Factor|Neural-restrictive silencer factor|Neuron Restrictive Silencer Factor|NRSF|RE1 silencing transcription factor|RE1-silencing transcription factor|Repressor binding to the X2 box|rest|REST protein|REST_HUMAN|X2 box repressor|XBR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13127
- Q13127
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice