Anti-CHST11 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CHST11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-874
- 77840-875
- Antigen Symbol:
- CHST11
- CHST11
- Antigen Name:
- Carbohydrate sulfotransferase 11 protein (214-330AA)
- Carbohydrate sulfotransferase 11 protein (214-330AA)
- Antigen Synonyms:
- C4S-1|C4ST 1|C4ST|C4ST-1|C4ST1|Carbohydrate (chondroitin 4) sulfotransferase 11|Carbohydrate sulfotransferase 11|Chondroitin 4 O sulfotransferase 1|Chondroitin 4 sulfotransferase|Chondroitin 4-O-sulfotransferase 1|Chondroitin 4-sulfotransferase 1|CHST 11|Chst11|CHSTB_HUMAN|HSA269537|IgH/CHST11 fusion
- C4S-1|C4ST 1|C4ST|C4ST-1|C4ST1|Carbohydrate (chondroitin 4) sulfotransferase 11|Carbohydrate sulfotransferase 11|Chondroitin 4 O sulfotransferase 1|Chondroitin 4 sulfotransferase|Chondroitin 4-O-sulfotransferase 1|Chondroitin 4-sulfotransferase 1|CHST 11|Chst11|CHSTB_HUMAN|HSA269537|IgH/CHST11 fusion
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NPF2
- Q9NPF2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice