Anti-MOCS1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: MOCS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-011
- 77840-012
- Antigen Symbol:
- MOCS1
- MOCS1
- Antigen Name:
- Molybdenum cofactor biosynthesis protein 1 protein (90-360AA)
- Molybdenum cofactor biosynthesis protein 1 protein (90-360AA)
- Antigen Synonyms:
- Cell migration-inducing gene 11 protein|MIG11|Migration inducing gene 11 protein|MOCOD|MOCODA|Mocs1|MOCS1_HUMAN|Molybdenum cofactor biosynthesis protein 1|Molybdenum cofactor biosynthesis protein A|Molybdenum cofactor biosynthesis protein C|Molybdenum cofactor synthesis 1|Molybdenum cofactor synthesis-step 1 protein A-B
- Cell migration-inducing gene 11 protein|MIG11|Migration inducing gene 11 protein|MOCOD|MOCODA|Mocs1|MOCS1_HUMAN|Molybdenum cofactor biosynthesis protein 1|Molybdenum cofactor biosynthesis protein A|Molybdenum cofactor biosynthesis protein C|Molybdenum cofactor synthesis 1|Molybdenum cofactor synthesis-step 1 protein A-B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZB8
- Q9NZB8
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice