Anti-G3BP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: G3BP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-847
- 77821-846
- Antigen Symbol:
- G3BP1
- G3BP1
- Antigen Name:
- Ras GTPase-activating protein-binding protein 1 protein (227-466AA)
- Ras GTPase-activating protein-binding protein 1 protein (227-466AA)
- Antigen Synonyms:
- ATP dependent DNA helicase VIII|ATP-dependent DNA helicase VIII|G3BP|G3BP stress granule assembly factor 1|G3BP-1|G3BP1_HUMAN|GAP binding protein|GAP SH3 domain binding protein 1|GAP SH3 domain-binding protein 1|GTPase activating protein (SH3 domain) binding protein 1|hDH VIII|Human DNA helicase VIII|MGC111040|Ras GTPase activating protein binding protein 1|Ras GTPase activating protein SH3 domain binding protein|Ras GTPase-activating protein-binding protein 1|RasGAP associated endoribonuclease G3BP
- ATP dependent DNA helicase VIII|ATP-dependent DNA helicase VIII|G3BP|G3BP stress granule assembly factor 1|G3BP-1|G3BP1_HUMAN|GAP binding protein|GAP SH3 domain binding protein 1|GAP SH3 domain-binding protein 1|GTPase activating protein (SH3 domain) binding protein 1|hDH VIII|Human DNA helicase VIII|MGC111040|Ras GTPase activating protein binding protein 1|Ras GTPase activating protein SH3 domain binding protein|Ras GTPase-activating protein-binding protein 1|RasGAP associated endoribonuclease G3BP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13283
- Q13283
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice