Anti-CCDC32 Rabbit Recombinant Polyclonal Antibody
Catalog # 77841-701
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Coiled-coil domain-containing protein 32 protein (1-185AA)
- Antigen Symbol:CCDC32
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BV29
- Antigen Synonyms:CCDC32| C15orf57Coiled-coil domain-containing protein 32
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: CCDC32
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human