Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-USP17L5 Rabbit Recombinant Polyclonal Antibody (FITC)
Anti-USP17L5 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-USP17L5 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77818-251
  • 77818-250
  • Antigen Symbol:
  • USP17L5
  • USP17L5
  • Antigen Name:
  • Ubiquitin carboxyl-terminal hydrolase 17-like protein 5 protein (19-235AA)
  • Ubiquitin carboxyl-terminal hydrolase 17-like protein 5 protein (19-235AA)
  • Antigen Synonyms:
  • Ubiquitin carboxyl-terminal hydrolase 17-like protein 5|EC 3.4.19.12|Deubiquitinating enzyme 17-like protein 5|Ubiquitin thioesterase 17-like protein 5|Ubiquitin-specific-processing protease 17-like protein 5
  • Ubiquitin carboxyl-terminal hydrolase 17-like protein 5|EC 3.4.19.12|Deubiquitinating enzyme 17-like protein 5|Ubiquitin thioesterase 17-like protein 5|Ubiquitin-specific-processing protease 17-like protein 5
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • A8MUK1
  • A8MUK1
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications