Anti-SUCLA2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: SUCLA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-740
- 77837-739
- Antigen Symbol:
- SUCLA2
- SUCLA2
- Antigen Name:
- Succinate--CoA ligase [ADP-forming] subunit beta, mitochondrial protein (1-180AA)
- Succinate--CoA ligase [ADP-forming] subunit beta, mitochondrial protein (1-180AA)
- Antigen Synonyms:
- A BETA|A SCS|ATP specific succinyl CoA synthetase subunit beta|ATP specific succinyl CoA synthetase, beta subunit|ATP-specific succinyl-CoA synthetase subunit beta|Mitochondrial succinyl CoA ligase [ADP forming] subunit beta|MTDPS5|Renal carcinoma antigen NY-REN-39|Renal carcinoma antigen NYREN39|SCS betaA|SCS-betaA|SUCB1_HUMAN|Succinate CoA ligase (ADP forming)|Succinate CoA ligase [ADP forming] subunit beta, mitochondrial, succinyl CoA ligase [ADP forming] subunit beta, mitochondrial|Succinate CoA ligase ADP forming beta subunit|Succinate CoA ligase beta subunit|Succinyl CoA ligase [ADP-forming] subunit beta, mitochondrial|Succinyl CoA synthetase beta A chain|Succinyl-CoA ligase [ADP-forming] subunit beta, mitochondrial|Succinyl-CoA synthetase beta-A chain|SUCLA2
- A BETA|A SCS|ATP specific succinyl CoA synthetase subunit beta|ATP specific succinyl CoA synthetase, beta subunit|ATP-specific succinyl-CoA synthetase subunit beta|Mitochondrial succinyl CoA ligase [ADP forming] subunit beta|MTDPS5|Renal carcinoma antigen NY-REN-39|Renal carcinoma antigen NYREN39|SCS betaA|SCS-betaA|SUCB1_HUMAN|Succinate CoA ligase (ADP forming)|Succinate CoA ligase [ADP forming] subunit beta, mitochondrial, succinyl CoA ligase [ADP forming] subunit beta, mitochondrial|Succinate CoA ligase ADP forming beta subunit|Succinate CoA ligase beta subunit|Succinyl CoA ligase [ADP-forming] subunit beta, mitochondrial|Succinyl CoA synthetase beta A chain|Succinyl-CoA ligase [ADP-forming] subunit beta, mitochondrial|Succinyl-CoA synthetase beta-A chain|SUCLA2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P2R7
- Q9P2R7
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice