Anti-USP12 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: USP12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77804-110
- 77804-109
- Antigen Symbol:
- USP12
- USP12
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 12 protein (1-370AA)
- Ubiquitin carboxyl-terminal hydrolase 12 protein (1-370AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 12|OTTHUMP00000042337|UBH 1|UBH1|Ubiquitin carboxyl terminal hydrolase 12|Ubiquitin carboxyl-terminal hydrolase 12|Ubiquitin hydrolyzing enzyme 1|Ubiquitin specific peptidase 12|Ubiquitin specific processing protease 12|Ubiquitin specific protease 12 like 1|Ubiquitin thioesterase 12|Ubiquitin thiolesterase 12|Ubiquitin-hydrolyzing enzyme 1|Ubiquitin-specific-processing protease 12|UBP12_HUMAN|USP 12|Usp12|USP12L1
- Deubiquitinating enzyme 12|OTTHUMP00000042337|UBH 1|UBH1|Ubiquitin carboxyl terminal hydrolase 12|Ubiquitin carboxyl-terminal hydrolase 12|Ubiquitin hydrolyzing enzyme 1|Ubiquitin specific peptidase 12|Ubiquitin specific processing protease 12|Ubiquitin specific protease 12 like 1|Ubiquitin thioesterase 12|Ubiquitin thiolesterase 12|Ubiquitin-hydrolyzing enzyme 1|Ubiquitin-specific-processing protease 12|UBP12_HUMAN|USP 12|Usp12|USP12L1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75317
- O75317
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together