Anti-PAPSS2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: PAPSS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77819-431
- 77819-430
- Antigen Symbol:
- PAPSS2
- PAPSS2
- Antigen Name:
- Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2 protein (2-138AA)
- Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2 protein (2-138AA)
- Antigen Synonyms:
- PAPSS2|ATPSK2Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2|PAPS synthase 2|PAPSS 2|Sulfurylase kinase 2|SK 2|SK2) [Includes: Sulfate adenylyltransferase|EC 2.7.7.4|ATP-sulfurylase|Sulfate adenylate transferase|SAT)|Adenylyl-sulfate kinase|EC 2.7.1.25|3'-phosphoadenosine-5'-phosphosulfate synthase|APS kinase|Adenosine-5'-phosphosulfate 3'-phosphotransferase|Adenylylsulfate 3'-phosphotransferase)]
- PAPSS2|ATPSK2Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2|PAPS synthase 2|PAPSS 2|Sulfurylase kinase 2|SK 2|SK2) [Includes: Sulfate adenylyltransferase|EC 2.7.7.4|ATP-sulfurylase|Sulfate adenylate transferase|SAT)|Adenylyl-sulfate kinase|EC 2.7.1.25|3'-phosphoadenosine-5'-phosphosulfate synthase|APS kinase|Adenosine-5'-phosphosulfate 3'-phosphotransferase|Adenylylsulfate 3'-phosphotransferase)]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95340
- O95340
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice