Anti-USP14 Rabbit Recombinant Polyclonal Antibody
77804-113
New Product
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 14 protein (1-494AA)
- Antigen Symbol:USP14
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P54578
- Antigen Synonyms:Deubiquitinating enzyme 14|TGT|tRNA guanine transglycosylase 60 kD subunit|tRNA guanine transglycosylase|Ubiquitin carboxyl terminal hydrolase 14|Ubiquitin carboxyl-terminal hydrolase 14|Ubiquitin specific peptidase 14|Ubiquitin specific processing protease 14|Ubiquitin specific protease 14|Ubiquitin thiolesterase 14|Ubiquitin-specific-processing protease 14|UBP14_HUMAN|USP 14|USP14
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: USP14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse