Anti-EXOSC5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: EXOSC5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77839-760
- 77839-761
- Antigen Symbol:
- EXOSC5
- EXOSC5
- Antigen Name:
- Exosome complex component RRP46 protein (1-235AA)
- Exosome complex component RRP46 protein (1-235AA)
- Antigen Synonyms:
- Chronic myelogenous leukemia tumor antigen 28|CML28|EXOS5_HUMAN|EXOSC 5|Exosc5|Exosome complex component rrp46|exosome complex exonuclease RRP46|Exosome component 5|exosome component Rrp46|hRrp46p|p12B|Ribosomal RNA processing protein 46|Ribosomal RNA processing protein 46, S. cerevisiae, homolog of|Ribosomal RNA-processing protein 46|RRP41B|RRP46|Rrp46p
- Chronic myelogenous leukemia tumor antigen 28|CML28|EXOS5_HUMAN|EXOSC 5|Exosc5|Exosome complex component rrp46|exosome complex exonuclease RRP46|Exosome component 5|exosome component Rrp46|hRrp46p|p12B|Ribosomal RNA processing protein 46|Ribosomal RNA processing protein 46, S. cerevisiae, homolog of|Ribosomal RNA-processing protein 46|RRP41B|RRP46|Rrp46p
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQT4
- Q9NQT4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice