Anti-TYRP1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TYRP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-790
- 77803-790
- Antigen Symbol:
- TYRP1
- TYRP1
- Antigen Name:
- 5,6-dihydroxyindole-2-carboxylic acid oxidase protein (91-186AA)
- 5,6-dihydroxyindole-2-carboxylic acid oxidase protein (91-186AA)
- Antigen Synonyms:
- 5|5,6 dihydroxyindole 2 carboxylic acid oxidase|6-dihydroxyindole-2-carboxylic acid oxidase|b-PROTEIN|CAS2|Catalase B|CATB|DHICA oxidase|Glycoprotein 75|GP75|Melanoma antigen gp75|OCA3|TRP|TRP-1|TRP1|Tyrosinase related protein 1|Tyrosinase-related protein 1|TYRP|TYRP1_HUMAN|TYRRP
- 5|5,6 dihydroxyindole 2 carboxylic acid oxidase|6-dihydroxyindole-2-carboxylic acid oxidase|b-PROTEIN|CAS2|Catalase B|CATB|DHICA oxidase|Glycoprotein 75|GP75|Melanoma antigen gp75|OCA3|TRP|TRP-1|TRP1|Tyrosinase related protein 1|Tyrosinase-related protein 1|TYRP|TYRP1_HUMAN|TYRRP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17643
- P17643
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together